Order Entry
Austria
ContactUsLinkComponent
Test kit, chloride test, Bresle method
Test kit, chloride test, Bresle method
Catalog # SIMESP7310
Supplier:  Industrial Physics
CAS Number:  
undefined
Test kit, chloride test, Bresle method
Catalog # SIMESP7310
Supplier:  Industrial Physics
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    TQC Bresle kit, for chloride ions
  • Seq:
    01
  • Model:
    Bresle Kit
  • Accuracy:
    1% of guage
  • Automatic switch-off:
    Yes
  • Calibration:
    at 84 µS/cm (autocalibration)
  • Compliance:
    IP 67 ( meter)
  • Measurement range:
    0,1 - 20000 mg/m² ( with DSP method)
  • Operating temperature:
    0...50 °C
  • Resolution:
    0,1 mg/m2 (with DSP method)
  • Range:
    0,1 - 20 000 mg/m2

Specifications

About this item

TQC Bresle kit is a chloride test kit that complies with the ISO 8502-6 and ISO 8502-9 standards. The Bresle method is designed to to assess the level of soluble salts using a Bresle patch or Bresle sampler, distilled water and a conductivity meter. The conductivity is mainly directly proportional to the concentration of dissolved chloride ions in the solution. The kit includes all the necessary equipment to execute a Bresle test that will indicate the contamination of soluble salts on blast-cleaned surfaces prior to coating. Included is a conductivity gauge used for the assessment of soluble salt ions as chlorides, sulphates and nitrates.

  • TQC DSP-procedure eliminates user-errors and saves time
  • Compact and complete kit in sturdy case
  • Unique Direct Sampling Procedure (DSP) to ensure high speed and accuracy
  • Up to 60 times more accurate than other test kits available

Contamination of blast media

It is also suitable to determine the contamination of blast-media in use. This important test described in the ISO 11127-6 and ISO 11127-7 standards helps to prevent the dissolved salts in the recycled abrasive media or water re-contaminating the surface being cleaned.

Normalisation temperature: 25 °C

Delivery: Kit supplied with digital conductivity meter, 25×Bresle patch, distilled water, cups and syringes, calibration and cleansing solutions, manual ( pictorial) and case.