Order Entry
Austria
ContactUsLinkComponent
Melting point and boiling point capillaries
Melting point and boiling point capillaries
Catalog # HILG2103037
Supplier:  HILGENBERG
undefined
Melting point and boiling point capillaries
Catalog # HILG2103037
Supplier:  HILGENBERG

Specifications

  • Ext.Ø:
    2,00 mm
  • Ø int.:
    1,60 mm
  • Length:
    100 mm
  • Environmentally Preferable:
  • Pk:
    1.000

Specifications

About this item

Soda lime glass, one end closed.