Order Entry
Austria
ContactUsLinkComponent
Combined ion selective electrodes, 500 series
Artikelnr. 662-0381
Lieferant:  WTW
undefined
Combined ion selective electrodes, 500 series
Artikelnr. 662-0381
Lieferant:  WTW

Spezifikationen

  • Concentration:
    0,02 - 900 mg/l (10-6...5 × 10-2 mol/l)
  • Connection:
    S7
  • Type:
    NH 500/2, gas-sensitive electrode for ammonium (NH4+)
  • Description:
    NH 500/2, gas-sensitive electrode, with integrated reference electrode
  • Seq:
    17
  • Pk:
    1
  • pH:
    4 - 12

Spezifikationen

Über diesen Artikel

These 500 series half cells require a separate reference electrode type R 503 (an exception for this is the gas-sensitive NH 500/2 with integrated reference electrode).

  • Ideal for challenging ISE applications
Ordering information: Reference electrodes, ionic strength adapters and standard solutions are not included. Please contact Avantor for further information.