Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Ammonium sulphide 20 - 24% in aqueous solution, AR®, Macron Fine Chemicals™
  3524-04
 :  
undefined
 
 
 
Ammonium sulphide 20 - 24% in aqueous solution, AR®, Macron Fine Chemicals™
  3524-04
 :  

 

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • For Laboratory,Research,or Manufacturing Use:
    20.0 - 24.0%
  • Assay (as (NH₄)₂S):
    20.0 - 24.0 %
  • Antimony (Sb):
    ≤0.0005 %
  • Arsenic (As):
    ≤0.0001 %
  • Carbonate (CO₃):
    ≤0.005 %
  • Chloride (Cl):
    ≤0.005 %
  • Residue after Ignition:
    ≤0.04 %

 

 

 :
 :68.14 g/mol
 :1.00 g/cm³
 :235-223-4
 :2683
 :8 (6.1) (3),II
 :GHS02
 :GHS05
 :GHS09
 :12135-76-1
 :H₈N₂S
 :MFCD00010892