CytoScan™-SRB cytotoxicity assay
Catalog # 786-213
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1.000 Assays
- Description:CytoScan™-SRB Cytotoxicity Assay Kit
- Tests per kit:1000 assays
Specifications
About this item
CytoScan™-SRB Cytotoxicity Assay is based upon the quantitative staining of cellular proteins by sulforhodamine B (SRB). The assay is used for cell density determination, based on the measurement of cellular protein content.
- Simple, accurate and reproducible assay
- Measures total biomass by staining cellular proteins
- Linear response
- For the detection of cell toxicity, death, viability or proliferation
- Ideal for high throughput screening