Orders LinkContactUsLinkComponent
VWR® Sample Vials
VWR® Sample Vials
Supplier:   VWR Collection
VWR® Sample Vials
Supplier:  VWR Collection
Certificates
VWR® Sample Vials
VWR® Sample Vials
2 Options Available Below
Product Details & Documents
These sample vials are made from PTFE.
  • Ideal for storing aggressive or expensive materials
  • Suitable for small scale reactions at low pressure
  • Tapered inner for easy removal of contents
  • Thick wall construction
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Availability
    Price
  • 25,00 ml
    33×64 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 50,00 ml
    43×78 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 136,00 €
    136,00 €
    1 * 1 ST
       136,00 €