Order Entry
Austria
ContactUsLinkComponent
Ammonia solution 27.0 - 30.0% NF, Multi-Compendial, J.T.Baker®
Corrosive
Irritant
Environment
Ammonia solution 27.0 - 30.0% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Environment
Ammonia solution 27.0 - 30.0% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
  • Meets N.F. Requirements:
  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Nonvolatile Residue:
    ≤0.05 %
  • NF - Readily Oxidizable Substances:
    Passes Test
  • NF - Assay (NH₃)(NF):
    27.0 - 31.0 %
  • EP/BP - Identification A (EP/BP):
    Passes Test
  • EP/BP - Identification B (EP/BP):
    Passes Test
  • EP/BP - Identification C (EP/BP):
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Oxidizable Substances:
    Passes Test
  • EP/BP - Pyridine and Related Substances:
    ≤2 ppm
  • EP/BP - Residue on Evaporation (mg/L):
    ≤20
  • EP/BP - Carbonates (CO₃):
    ≤60 ppm
  • EP/BP - Chlorides (Cl):
    ≤1 ppm
  • EP/BP - Sulfates (SO₄):
    ≤5 ppm
  • EP/BP - Iron (Fe):
    ≤0.25 ppm
  • EP/BP - Assay (NH₃)(EP/BP):
    25.0 - 30.0 %

Specification Test Results

Related Information

Frequently Bought Together