ContactUsLinkComponent
VWR® Microban®, Glove Box Holders
VWR® Microban®, Glove Box Holders
Catalog # 211-0371
Supplier:  VWR Collection
undefined
VWR® Microban®, Glove Box Holders
Catalog # 211-0371
Supplier:  VWR Collection
Specifications
  • Colour:
    Blue
  • Description:
    Glove box holders, Microban®
  • W×D×H:
    254×156×97 mm
  • Pk:
    3
Specifications
About this item
These wall-mountable glove box holders are made from polystyrene with Microban® additive.
  • Microban® additive prevents growth of moulds and bacteria
  • High impact polystyrene, resistant to harsh cleaning solutions
  • Modular design allows stacking to save space
Suitable for use in labs, cleanrooms, hospitals and microbiological work areas.
Delivery: Mounting screws included.