Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Ammonium thiocyanate 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5627-02
Supplier:  Avantor
CAS Number:  
undefined
Ammonium thiocyanate 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5627-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.095 - 0.105
  • Normality:
    0.095 - 0.105

Specification Test Results

About this item

 :
 :76.12 g/mol
Density:1.10 g/cm³
Storage Temperature:Ambient
EINECS:217-175-6
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428