ContactUsLinkComponent
Immobilised Pepsin
Immobilised Pepsin
Catalog # 786-791
Supplier:  G-Biosciences
CAS Number:  
undefined
Immobilised Pepsin
Catalog # 786-791
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Pepsin, immobilised
Specifications
About this item
A proteolytic enzyme immobilised on 4% agarose that is routinely used for the generation of F(ab’)₂ fragments from Immunoglobulin G (IgG). The pepsin has the ability to cleave the heavy chains near the hinge region. The immobilised Pepsin offers the distinct advantage of eliminating enzyme contamination of the F(ab)₂ fragments.
  • Generate F(ab)₂ fragments
  • Eliminate contaminating pepsin enzyme
  • Can be used in virtually all scenarios using free pepsin