Rat single tissue blot, single species
Supplier: G-Biosciences
Certificates
About this item
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
- Ready to screen
- Single tissue rat blots
Frequently Bought Together