Order Entry
Austria
Orders LinkContactUsLinkComponent
GET™ Total RNA kit

GET™ Total RNA kit

GET™ Total RNA Kit, 50×50 ml preps, 1 KIT
Catalog # 786-132
Supplier:   G-Biosciences
GET™ Total RNA kit
Catalog # 786-132
Supplier:  G-Biosciences
CAS Number:  
GET™ Total RNA kit
GET™ Total RNA kit

GET™ Total RNA Kit, 50×50 ml preps, 1 KIT

CAS Number:  
Price
389,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

The GET™ Total RNA Kit isolates total RNA from contaminating DNA, proteins, and nucleases using our GET™ RNA spin columns. After homogenisation, RNA is bound to the GET™ RNA spin columns and washed. The eluted RNA is ready for any procedure including Northern/slot/dot blots, reverse transcription or RNase protective assays.

  • Rapidly inhibit destructive RNases
  • Strong chaotropic buffer for rapid RNA release
  • High affinity RNA binding spin columns
  • Optional DNase treatment
  • Compatible with numerous species and tissues

Documents

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    GET™ Total RNA Kit, 50×50 ml preps
  • Cat. No.:
    786-132
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)|Organic extraction
  • Description:
    GET™ Total RNA Kit
  • Sample type:
    Plant samples|Bacteria|Cultured cells|Human/animal cells and tissues

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.