Order Entry
Austria
ContactUsLinkComponent
RNA purification kit, Tri-Xtract™
RNA purification kit, Tri-Xtract™
Supplier:  G-Biosciences
Certificates
undefined
RNA purification kit, Tri-Xtract™
Supplier:  G-Biosciences

Specifications

  • Method / Format:
    Precipitation
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
    Total DNA
    Total protein
  • Sample type:
    Plant samples
    Bacteria
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
    RNA
    Protein
    Co-purification of RNA, DNA and protein
  • Description:
    Tri-Xtract™

Specifications

Related Information

Frequently Bought Together