GET™ Agarose DNA Kit
GET™ AGAROSE DNA method involves release of DNA fragments from gel pieces followed by capture of nucleic acids on the GET™ spin columns, washing, and elution of the clean nucleic acid fragments in a suitable buffer.
- Extract DNA fragments from agarose gels
- Rapid DNA isolation
- DNA ready for downstream applications, including ligations
- Suitable for 100 to 20000 bp fragments
- Compatible with TAE and TBE buffer gels
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:DNA gel bands
- Sample type:Gel matrix
- Target molecule:DNA
- Description:GET™ AGAROSE DNA Kit
Frequently Bought Together