Order Entry
Austria
ContactUsLinkComponent
GET™ CLEAN DNA Kit
GET™ CLEAN DNA Kit
Supplier:  G-Biosciences
Certificates
undefined
GET™ CLEAN DNA Kit
Supplier:  G-Biosciences

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    GET™ CLEAN DNA Kit

Specifications

Related Information

Frequently Bought Together