Order Entry
Austria
Orders LinkContactUsLinkComponent
Crates, stackable

Crates, stackable

Supplier:   LAMAPLAST
Crates, stackable
Supplier:  LAMAPLAST
Certificates
Crates, stackable
Crates, stackable

2 Options Available Below

Product Details & Documents

HDPE, white.

  • Temperature resistance : −40 to 80 °C
  • Washing 120 °C for food
  • Closed handle

The stackable crates with closed bottom and walls are ideal for food use.

Product Family Options

Product Information

  • Capacity
    W×D×H ext.
    W×D×H int.
    Availability
    Price
  • 10 l
    400×300×100 mm
    365×265×95 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €

  • 12 l
    600×400×70 mm
    565×365×65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,50 €
    45,50 €
    1 * 1 ST
       45,50 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.