Order Entry
Austria
ContactUsLinkComponent
Yeast Geno-DNA Template™
Yeast Geno-DNA Template™
Catalog # 786-134
Supplier:  G-Biosciences
CAS Number:  
undefined
Yeast Geno-DNA Template™
Catalog # 786-134
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    Yeast Geno-DNA-Template™, 100 tests
  • Description:
    Yeast Geno-DNA-Template™

Specifications

About this item

Yeast Geno-DNA Template™ isolates high quality genomic DNA from yeast. The kit is based on a two-step lysis of yeast cells, using LongLife™ Zymolyase® and LongLife™ Proteinase K, followed by the removal of proteins and other cellular impurities by precipitation and centrifugation.

The genomic DNA is collected by precipitation and the high yield of extracted DNA has an average 100 kb length and has A₂₆₀/A₂₈₀ 1,8 to 2,0.