Orders LinkContactUsLinkComponent
Grindometers
Grindometers
Supplier:   Industrial Physics
Grindometers
Supplier:  Industrial Physics
Certificates
Grindometers
11 Options Available Below
Product Details & Documents
These grindometers or gauges of fineness are precision instruments to determine particle size and fineness of many materials like paints, lacquers, pigments, filler, chocolate and more. TQC Grindometers have double grooves with graded slopes graduated in three different parameters: µm (microns), NS (Hegman) and PCU (North).
  • Gauge and beveled scraper are made of hardened stainless steel and have an accuracy of 2 µm
W×D×H: 175×60×12 mm
Certifications: Conforms to DIN EN ISO NF21524.
Delivery information: Supplied with test certificate.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • SIMEVF2110
    Grindometer, double groove, 0 - 15 µm, graduation 1,5 µm
    Grindometer, double groove, 0 - 15 µm, graduation 1,5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 536,00 €
    536,00 €
    1 * 1 ST
       536,00 €
  • SIMEVF2120
    Grindometer, wide groove, 0 - 15 µm, graduation 1,5 µm
    Grindometer, wide groove, 0 - 15 µm, graduation 1,5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
  • SIMEVF2111
    Grindometer, double groove, 0 - 25 µm, graduation 2,5 µm
    Grindometer, double groove, 0 - 25 µm, graduation 2,5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 536,00 €
    536,00 €
    1 * 1 ST
       536,00 €
  • SIMEVF2121
    Grindometer, wide groove, 0 - 25 µm, graduation 2,5 µm
    Grindometer, wide groove, 0 - 25 µm, graduation 2,5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
  • SIMEVF2112
    Grindometer double groove, 0 - 50 µm, graduation 5 µm
    Grindometer double groove, 0 - 50 µm, graduation 5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 536,00 €
    536,00 €
    1 * 1 ST
       536,00 €
  • SIMEVF2122
    Grindometer, wide groove, 0 - 50 µm, graduation 5 µm
    Grindometer, wide groove, 0 - 50 µm, graduation 5 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
  • SIMEVF2113
    Grindometer, double groove, 0 - 100 µm, graduation 10 µm
    Grindometer, double groove, 0 - 100 µm, graduation 10 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 536,00 €
    536,00 €
    1 * 1 ST
       536,00 €
  • SIMEVF2123
    Grindometer, wide groove, 0 - 100 µm, graduation 10 µm
    Grindometer, wide groove, 0 - 100 µm, graduation 10 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €
  • SIMEVF2108
    Grindometer, single groove, 0 - 250 µm, graduation 25 µm
    Grindometer, single groove, 0 - 250 µm, graduation 25 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 779,00 €
    779,00 €
    1 * 1 ST
       779,00 €
  • SIMEVF2118
    Grindometer, double groove, 0 - 250 µm, graduation 25 µm
    Grindometer, double groove, 0 - 250 µm, graduation 25 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 696,00 €
    696,00 €
    1 * 1 ST
       696,00 €
  • SIMEVF2124
    Grindometer, double groove, 0 - 500 µm, graduation 50 µm
    Grindometer, double groove, 0 - 500 µm, graduation 50 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 844,00 €
    844,00 €
    1 * 1 ST
       844,00 €