Genomic DNA isolation and purification kits, GET™ DNA Template
Supplier: G-Biosciences
Certificates
About this item
GET™ DNA Template is a spin column format kit suitable for the preparation of DNA templates from blood, cells, fungal and animal tissue samples. The isolated genomic DNA is suitable for PCR and other downstream applications.
- Spin column format
- Purifies mammalian genomic DNA free from impurities
- Yield is ~1 to 40 µg DNA per preparation
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:BloodYeast/FungiCultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Description:GET™ DNA Template Kit
Specifications
Frequently Bought Together