Genomic DNA isolation From mouse tail, OmniPrep™
Catalog # 786-401
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 KIT
- Method / Format:PrecipitationOrganic extraction
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:DNA
- Description:OmniPrep™ for Mouse Tail
- Description:OmniPrep™ for Mouse Tail
- Yield:70 - 80 μg/cm tail
Specifications
About this item
OmniPrep™ for Mouse Tail Kit isolates high quality genomic DNA from mouse tail samples. If used according to the protocols this kit purifies DNA from 100 to 200 cm mouse tail.
- High Yield, ~100 kb genomic DNA
- A₂₆₀/A₂₈₀ between 1,8 and 2,0
- Simple two tube method, 20 to 40 minutes
- Supplied with extra LongLife™ Proteinase K for efficient DNA release
- No toxic chemicals, no phenol, no hazardous waste