Genomic DNA isolation from fungi, OmniPrep™ for fungi kit
Catalog # 786-399
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 KIT
- Method / Format:PrecipitationOrganic extraction
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Yeast/Fungi
- Target molecule:DNA
- Description:OmniPrep™ for Fungi
- Description:OmniPrep™ for Fungi
- Yield:0.2 - 1 µg/5 mg fungal samples
Specifications
About this item
OmniPrep™ for Fungi Kit isolates high quality genomic DNA from fungal samples.If used according to the protocols this kit purifies DNA from 1 to 2 g fungal tissues.
- High Yield, ~100 kb genomic DNA
- A₂₆₀/A₂₈₀ between 1, 8 and 2,0
- Simple two tube method, 20 to 40 minutes
- Molecular Grinding Resin™ for efficient fungal disruption
- No toxic chemicals, no phenol, no hazardous waste