Order Entry
Austria
ContactUsLinkComponent
Genomic DNA isolation from bacteria, OmniPrep™ for Gram Positive Bacteria kit
Genomic DNA isolation from bacteria, OmniPrep™ for Gram Positive Bacteria kit
Catalog # 786-398
Supplier:  G-Biosciences
CAS Number:  
undefined
Genomic DNA isolation from bacteria, OmniPrep™ for Gram Positive Bacteria kit
Catalog # 786-398
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
    Organic extraction
  • Prep size:
    Scalable
  • Refine target molecule:
    Bacterial DNA
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Description:
    OmniPrep™ for Gram Positive Bacteria
  • Description:
    OmniPrep™ for Gram Positive Bacteria
  • Yield:
    25 - 50 μg/ml Gram-positive culture

Specifications

About this item

OmniPrep™ for Gram Positive Bacteria Kit isolates high quality genomic DNA from gram-positive bacteria. If used according to the protocols, this kit purifies DNA from 100 ml gram-positive bacteria culture.

  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Simple two tube method, 20 to 40 minutes
  • No toxic chemicals, no phenol, no hazardous waste
  • Supplied with LongLife™ Lysozyme for rapid bacterial cell wall digestion