Cable ties, non-releasable, Allplastik-Kabelbinder®
Supplier: WUERTEMBERGISCHE ALLPLASTIK
Certificates
About this item
These Allplastik-Kabelbinder® cable ties are made of high-quality polyamide 6.6, have high tensile strength and are available in different lengths.
- With a curved round tip to make it easier to thread into the eye of the cable tie
- Rounded edges for safe usage
- Material with minimal coefficients of friction
- Printing or laser engraving is possible
- Temperature resistance: −40 to +85 °C
The ties are ideal for electrical installations, for wiring in industrial systems and control boards, for the automobile sector.
Certifications: IEC EN 62275.
Frequently Bought Together