Order Entry
Austria
ContactUsLinkComponent
 
Genomic DNA isolation and purification kits, OmniPrep™
Catalog # 786-136
Supplier:  G-Biosciences
CAS Number:  
undefined
Genomic DNA isolation and purification kits, OmniPrep™
Catalog # 786-136
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    OmniPrep™, 150 preps
  • Description:
    OmniPrep™

Specifications

About this item

The OmniPrep™ kit isolates high quality genomic DNA from many different species and tissue types including animal, plant, bacteria, yeast, fungi, whole blood, and cells in culture. DNA can be isolated from samples high in polysaccharides or other contaminants that are difficult to remove from the DNA preparations.

  • High Yield, ~100 kb genomic DNA
  • A₂₆₀/A₂₈₀ between 1,8 and 2,0
  • Adaptable for variety of samples, including plant, bacteria, fungi, mammalian tissues, cells and blood
  • Simple two tube method, 20 to 40 minutes
  • No toxic chemicals, no phenol, no hazardous waste
  • Suitable for ~150 preparations, depending on sample