Order Entry
Austria
ContactUsLinkComponent
Sample cups for immunoassay analysers
Sample cups for immunoassay analysers
Catalog # SARS73.663
Supplier:  SARSTEDT
undefined
Sample cups for immunoassay analysers
Catalog # SARS73.663
Supplier:  SARSTEDT

Specifications

  • Ext.Ø:
    12 mm
  • Capacity:
    2 ml
  • Colour:
    Transparent
  • Description:
    For immuno analyser ES 600 and ES 300
  • Height:
    40 mm
  • Pk:
    5.000

Specifications

About this item

PS, transparent.

Disposable cups for serum samples, immuno analysers ES 600 and ES 300.