Order Entry
Austria
ContactUsLinkComponent
Fragment selection kits, Axygen® AxyPrep™ FragmentSelect-I Kit
Fragment selection kits, Axygen® AxyPrep™ FragmentSelect-I Kit
Catalog # MAG-FRAG-I-5
Supplier:  Corning
CAS Number:  
Fragment selection kits, Axygen® AxyPrep™ FragmentSelect-I Kit
Catalog # MAG-FRAG-I-5
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    AxyPrep™ FragmentSelect-I Kit
  • Volume:
    5 ml
  • No. of reactions:
    75-140 preps

Specifications

About this item

The AxyPrep™ FragmentSelect Kits utilise a paramagnetic bead technology for quick high throughput optimised DNA size selection suitable for various next generation sequencing platforms. The AxyPrep™ FragmentSelect-I kit is optimised for Illumina next generation sequencing platforms and Life Technologies (SOLiD™) fragment size selection needs simplifying the library construction workflow.

  • Optimized fragment selection chemistries for Illumina, SOLiD, and Roche/454
  • Versatile and flexible sequential fragment size selection; no gels needed
  • Efficient removal of primer-dimers; no need for further clean-up steps
  • Manual and automation friendly
  • Scalable: Tube, 96-, and 384-well plate formats
  • No centrifugation or filtration required

Post DNA shearing, the library construction process for next generation sequencing mostly requires fragment selection regardless of the platform.

The AxyPrep™ FragmentSelect-I Kits utilize a unique paramagnetic bead technology for quick high-throughput optimized DNA size selection suitable for various next generation sequencing platforms.

Obtaining high recovery post fragment selection is becoming an important contributor for the reduction of sequencing bias.