Order Entry
Austria
ContactUsLinkComponent
Temperature probes
Temperature probes
Catalog # 454-3050
Supplier:  ISOPAD
Temperature probes
Catalog # 454-3050
Supplier:  ISOPAD

Specifications

  • Range:
    −80...+250 °C
  • Description:
    Temperature sensor, TAI-PM, Pt 100, sheathed element, with PTFE connecting bend
  • Length:
    2,0 m (connecting bend)
  • Pk:
    1
  • Ø×L:
    3,5×50 mm

Specifications

About this item