Orders LinkContactUsLinkComponent
Stereo zoom microscope
Eyepiece with dioptre adjuster and rubber eyeshield
Catalog # NIKOMMK30102
Supplier:   NIKON
Stereo zoom microscope
Catalog # NIKOMMK30102
Supplier:  NIKON
Stereo zoom microscope
Eyepiece with dioptre adjuster and rubber eyeshield
Price
116,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This stereo zoom microscope is designed to be used in environments where the temperature and humidity are high.
  • Ergonomic zooming knob with anti-mould design
  • Click-stop for each magnification
  • ESD protection to prevent samples from being damaged by electrostatic discharge
  • Greenough optical system, magnification from 3,35 to 300×, high optical zoom magnification 7,5×
  • Reflection prism for bright, high contrast images to be obtained
Specifications
  • Magnification:
    C-WF 10×/22
  • Description:
    Eyepiece with dioptre adjuster and rubber eyeshield
  • Seq:
    100