Order Entry
Austria
ContactUsLinkComponent
 
PCR clean-up kit, Axygen® AxyPrep™ Mag
 :  Corning
 
undefined
PCR clean-up kit, Axygen® AxyPrep™ Mag
 :  Corning
 

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    AxyPrep™ Mag PCR Clean Up Kit

 

Frequently Bought Together