Orders LinkContactUsLinkComponent
Fraction-FOCUS™
Fraction-FOCUS™, 10 Tests
Catalog # 786-168
Supplier:   G-Biosciences
Fraction-FOCUS™
Catalog # 786-168
Supplier:  G-Biosciences
CAS Number:  
Fraction-FOCUS™
Fraction-FOCUS™
Fraction-FOCUS™, 10 Tests
CAS Number:  
Price
736,00 €
Log in for availability
1 * 10 Tests
Product Details & Documents
Fraction-FOCUS™ utilises proven technology to fractionate and concentrate all proteomes into multiple fractions; simplifying 2D maps and enhancing detection of low abundant proteins. Fraction-FOCUS™ is fully compatible with all downstream protein identification techniques.
There is no detectable loss of total protein during the Fraction-FOCUS™ procedure. At the end of the fractionation, cellular proteins are in one of many fractions.
Documents
Specifications
Specifications
  • Pk:
    10 Tests
  • Description:
    Fraction-FOCUS™