ContactUsLinkComponent
Nucleic acid and protein destain, Silver D-Stain™
Nucleic acid and protein destain, Silver D-Stain™
Catalog # 786-199
Supplier:  G-Biosciences
CAS Number:  
undefined
Nucleic acid and protein destain, Silver D-Stain™
Catalog # 786-199
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    25
  • Description:
    Nucleic acid and protein destains, Silver D-Stain™
Specifications
About this item
The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stain™ eliminates the need to repeat the electrophoresis.
Suitable for reducing the staining of silver stained gels and revealing over-stained bands.