Orders LinkContactUsLinkComponent
Microbact™
Microbact™
Microbact™ GNB 24E, 40 Tests
Catalog # OXOIMB1131A
Microbact™
Catalog # OXOIMB1131A
CAS Number:  
Microbact™
Microbact™ GNB 24E, 40 Tests
CAS Number:  
Price
415,00 €
Log in for availability
1 * 40 Tests
Product Details & Documents
The Microbact™ Gram-negative system is to be used for the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram- negative bacteria). Organism identification is based on pH change and substrate utilisations as established by published reference methodologies.
  • Fast
  • Simple
  • Reliable
Specifications
Specifications
  • Pk:
    40 Tests
  • Description:
    Microbact™ GNB 24E
Specifications