Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Certificates
13 Options Available Below
Product Details & Documents
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • OXOIDS0147A
    TTC Red Spot
    TTC Red Spot
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 56,70 €
    56,70 €
    1 * 10 ST
       56,70 €
  • OXOIDS0155A
    TTC red spot /Malt extract agar
    TTC red spot /Malt extract agar
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 61,20 €
    61,20 €
    1 * 10 ST
       61,20 €
  • OXOIDS0166A
    PCA / MacConkey agar No.3
    PCA / MacConkey agar No.3
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 53,60 €
    53,60 €
    1 * 10 ST
       53,60 €
  • OXOIDS0167A
    PCA / MacConkey No.3 with germicide
    PCA / MacConkey No.3 with germicide
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 50,10 €
    50,10 €
    1 * 10 ST
       50,10 €
  • OXOIDS0168A
    PCA / VRBG agar
    PCA / VRBG agar
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,70 €
    59,70 €
    1 * 10 ST
       59,70 €
  • OXOIDS0169A
    PCA / VRBG agar with germicide
    PCA / VRBG agar with germicide
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 58,60 €
    58,60 €
    1 * 10 ST
       58,60 €
  • OXOIDS0170A
    PCA / Ogy agar
    PCA / Ogy agar
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 56,10 €
    56,10 €
    1 * 10 ST
       56,10 €
  • OXOIGFD04A
    TSA/Malt extract
    TSA/Malt extract
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 30,60 €
    30,60 €
    1 * 10 ST
       30,60 €
  • OXOIGFD02A
    CLED/MacConkey/Malt extract
    CLED/MacConkey/Malt extract
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 28,40 €
    28,40 €
    1 * 10 ST
       28,40 €
  • OXOIGFD03A
    CLED/MacConkey/Cetrimide
    CLED/MacConkey/Cetrimide
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 29,70 €
    29,70 €
    1 * 10 ST
       29,70 €
  • OXOIGFD05A
    PC/MacConkey+ MUG
    PC/MacConkey+ MUG
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
  • Plate Count/VRBG
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 30,90 €
    30,90 €
    1 * 10 ST
       30,90 €
  • OXOIGFD07A
    Plate Count/Plate Count
    Plate Count/Plate Count
    10
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 38,60 €
    38,60 €
    1 * 10 ST
       38,60 €