Order Entry
Austria
Orders LinkContactUsLinkComponent
Blocking solution, BLOK™ BSA

Blocking solution, BLOK™ BSA

BLOK™ BSA in TBS, 10% solution, 125 mL
Catalog # 786-193
Supplier:   G-Biosciences
Blocking solution, BLOK™ BSA
Catalog # 786-193
Supplier:  G-Biosciences
CAS Number:  
Blocking solution, BLOK™ BSA
Blocking solution, BLOK™ BSA

BLOK™ BSA in TBS, 10% solution, 125 mL

CAS Number:  
Price
241,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

BLOK™ BSA is a 10% BSA protein blocking solution.

  • For blocking Westerns, ELISA and dot blots
  • Ready-to-use
  • Available in TBS or PBS buffers

Documents

Specifications

Specifications

  • Pk:
    125 mL
  • Description:
    BLOK™ BSA in TBS, 10% solution

Specifications