Orders LinkContactUsLinkComponent
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
Blocking solution, BLOK™ Casein
BLOK™ Casein is a purified protein, used for blocking free binding sites in Western blot, ELISA plate well, immunohistochemistry and other immunochemical applications.
  • Ready to use
  • Available in the choices of TBS or PBS buffers
Product Family Options
  • Description
    Pk
  • BLOK™ Casein in PBS, 1% solution
    2
    1 * 2 ST
    1 * 2 ST - 209,00 €
    209,00 €
    1 * 2 ST
       209,00 €
  • BLOK™ Casein in TBS, 1% solution
    2
    1 * 2 ST
    1 * 2 ST - 184,00 €
    184,00 €
    1 * 2 ST
       184,00 €