ContactUsLinkComponent
Filter cards, brown, for TPX chambers, SHANDON™
Filter cards, brown, for TPX chambers, SHANDON™
Catalog # SHAN5991023
Supplier:  Epredia
undefined
Filter cards, brown, for TPX chambers, SHANDON™
Catalog # SHAN5991023
Supplier:  Epredia
Specifications
  • Description:
    Filter cards, brown
  • Pk:
    200
Specifications
About this item
Highly absorbent disposable filter cards for use with TPX chamber.
  • Use for sample volumes of 0,4 ml or less
  • For single chambers
Not for use with Cytofunnel™ disposable sample chambers.