ContactUsLinkComponent
Protein blocking agent, BLOT-QuickBlocker™
Protein blocking agent, BLOT-QuickBlocker™
Catalog # 786-011
Supplier:  G-Biosciences
CAS Number:  
undefined
Protein blocking agent, BLOT-QuickBlocker™
Catalog # 786-011
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    175 g
  • Description:
    BLOT-QuickBlocker™ blocking agent
Specifications
About this item
BLOT-QuickBlocker™ is a modified milk protein that is highly soluble and does not inhibit peroxidase detection. The modified milk protein has high blocking efficiency with a clear background.
  • Readily soluble and produces semi-clear solution
  • No inhibition to peroxidase
  • Fat free and produces clear background
  • Higher blocking efficiency
  • Blocking time 30 to 60 minutes