Orders LinkContactUsLinkComponent
Blocking buffers, FirstChoice™ | Avantor
Blocking buffers, FirstChoice™
Supplier:   G-Biosciences
Blocking buffers, FirstChoice™
Supplier:  G-Biosciences
Certificates
Blocking buffers, FirstChoice™
Blocking buffers, FirstChoice™
4 Options Available Below
Product Details & Documents
FirstChoice™ Blocking Buffer is ideal as a first choice for optimization of new assays and systems, or when determining the optimal blocking buffer for elimination of non specific binding sites in ELISA, blotting, immunohistochemistry and other applications. Buffers are conveniently supplied in widely used TBS (Tris-buffered saline at pH 7,5) and PBS (phosphate-buffered saline at pH 7,5) buffers as well as in separate formulations containing Tween® 20 for improving blocking efficiencies.
  • Ready to use
  • Greater versatility
  • Lack of cross reactivity
  • Compatible with antibodies and avidin/biotin based system
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 786-666
    FirstChoice™ blocking buffer-TBS
    FirstChoice™ blocking buffer-TBS
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 150,00 €
    150,00 €
    1 * 500 mL
       150,00 €
  • 786-667
    FirstChoice™ blocking buffer-TBST
    FirstChoice™ blocking buffer-TBST
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 158,00 €
    158,00 €
    1 * 500 mL
       158,00 €
  • 786-668
    FirstChoice™ blocking buffer-PBS
    FirstChoice™ blocking buffer-PBS
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 150,00 €
    150,00 €
    1 * 500 mL
       150,00 €
  • 786-669
    FirstChoice™ blocking buffer-PBST
    FirstChoice™ blocking buffer-PBST
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 158,00 €
    158,00 €
    1 * 500 mL
       158,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.