ContactUsLinkComponent
Glove liners, Puri ESD Top Fit
Glove liners, Puri ESD Top Fit
Supplier:  PURITECH
Certificates
undefined
Glove liners, Puri ESD Top Fit
Supplier:  PURITECH
Specifications
  • Catalog No:
  • 112-6221
  • 112-6218
  • 112-6219
  • 112-6222
  • 112-6220
  • Model:
  • Puri ESD Top Fit
  • Puri ESD Top Fit
  • Puri ESD Top Fit
  • Puri ESD Top Fit
  • Puri ESD Top Fit
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Anti-static:
  • +
  • +
  • +
  • +
  • +