Order Entry
Austria
ContactUsLinkComponent
SDS detection and estimation reagent kit
SDS detection and estimation reagent kit
Catalog # 786-129
Supplier:  G-Biosciences
CAS Number:  
undefined
SDS detection and estimation reagent kit
Catalog # 786-129
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    SDS Detection and Estimation Reagent Kit

Specifications

About this item

A reagent kit for detection and estimation of SDS in a sample. Mix the test sample in the extraction buffer reagents provided with the kit. If SDS is present in the sample, a blue colour is extracted that can be quantitatively measured.

Assay range: 0,02 to 0,2% SDS.