ContactUsLinkComponent
Protease inhibitor cocktail, Recom ProteaseArrest™
Protease inhibitor cocktail, Recom ProteaseArrest™
Catalog # 733-1997
Supplier:  G-Biosciences
CAS Number:  
undefined
Protease inhibitor cocktail, Recom ProteaseArrest™
Catalog # 733-1997
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    Recom ProteaseArrest™ (100X)
Specifications
About this item
Recom ProteaseArrest™ is a broad range, bacterial, 100X concentrated, ready to use protease inhibitor cocktail. Recom ProteaseArrest™ offers greater protection for recombinant proteins expressed and purified from bacteria. Recom ProteaseArrest™ inhibits bacterial serine, cysteine, metallo- and other bacterial specific proteases, including aminopeptidases and aspartic proteases.