General purpose gloves, AlphaTec® 87-195
Supplier: Ansell
Certificates
About this item
These natural rubber latex gloves with cotton flock lining provide liquid and mechanical resistance.
- Specially treated to reduce the risk of allergic reactions
- Straight cuff
- Provides outstanding sensitivity
- Fishscale design provides good grip
Ideal for use in food applications, such as poultry processing, beef and pig processing, fish processing and fruit and vegetable processing. Also suitable for janitorial use and cleaning applications.
Certifications: EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved
Food contact approved
Specifications
- Catalog No:
- 112-6437
- 112-6438
- 112-6439
- 112-6440
- Model:
- 87-195
- 87-195
- 87-195
- 87-195
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Fishscale
- Fishscale
- Fishscale
- Fishscale
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (1010X), EN ISO 374-1 Type B (KLT), EN ISO 374-5 VIRUS, EN 421
- Food contact approved:
- +
- +
- +
- +