ContactUsLinkComponent
Flasks, round bottom, with three necks and standard ground joints
Flasks, round bottom, with three necks and standard ground joints
Catalog # NEUB0759-0025-14
Flasks, round bottom, with three necks and standard ground joints
Catalog # NEUB0759-0025-14
Specifications
  • Capacity:
    25 ml
  • Pk:
    1
  • Side necks:
    14/23 NS
  • Centre neck:
    14/23 NS
Specifications
About this item
Borosilicate glass 3.3, clear, with angled side necks.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.