ContactUsLinkComponent
Syringe filters, Minisart® PES
Syringe filters, Minisart® PES
Supplier:  Sartorius
Certificates
undefined
Syringe filters, Minisart® PES
Supplier:  Sartorius
Specifications
  • Catalog No:
  • SART1757H-K
  • 516-0682
  • SART1757H-Q
  • SART1776D-Q
  • Airflow rate:
  • 1200 ml/min with air at 0,1 bar (1,5 psi)
  • 40 ml/min with water at 1 bar (14,5 psi)
  • 1200 ml/min with air at 0,1 bar (1,5 psi)
  • 40 ml/min with water at 1 bar (14,5 psi)
  • Max. operating pressure:
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • Max. temperature:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)