Solid phase microextraction fibers
About this item
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
31 Options Available Below
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Product Family Options
Septum inlet
Product Information
- DescriptionVerfügbarkeitPreis
- SPME fiber for autosampler use, carboxen/PDMS, 24 gauge, 75 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for autosampler use, PDMS/DVB, 24 gauge, 65 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for autosampler use, carboxen/PDMS, 85 µm fiber coating, StableflexLog in for availability1 * 3 ST
- SPME fiber for autosampler use, DVB/carboxen/PDMS, 50/30 µm fiber coating, StableflexLog in for availability1 * 3 ST
- SPME fiber for autosampler use, PDMS/DVB, 65 µm fiber coating, StableflexLog in for availability1 * 3 ST
- SPME fiber for manual use, carboxen/PDMS, 24 gauge, 75 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for manual use, DVB/carboxen/PDMS, 50/30 µm fiber coating, StableflexLog in for availability1 * 1 ST
- SPME fiber for manual use, DVB/carboxen/PDMS, 50/30 µm fiber coating, StableflexLog in for availability1 * 1 ST
Merlin microseal inlet
Product Information
- DescriptionVerfügbarkeitPreis
- SPME fiber for autosampler use, carbowax/PEG, 23 gauge, 60 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for autosampler use, carboxen/PDMS, 23 gauge, 75 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for autosampler use, PDMS, 23 gauge, 100 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for autosampler use, PDMS/DVB, 23 gauge, 65 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for manual use, carbowax/PEG, 23 gauge, 60 µm fiber coatingLog in for availability1 * 3 ST
- SPME fiber for manual use, carboxen/PDMS, 23 gauge, 75 µm fiber coatingLog in for availability1 * 3 ST
Kits
Product Information
- DescriptionVerfügbarkeitPreis
- Kit 1: polyacrylate, PDMS, PDMS, for volatiles and semivolatiles, for autosampler useLog in for availability1 * 3 ST
- Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for autosampler useLog in for availability1 * 1 ST
- Kit 4: PDMS, PDMS/DVB and carboxen/PDMS, for flavors and odors, for autosampler useLog in for availability1 * 1 ST
- StableFlex fiber kit: PDMS/DVB, DVB/carboxen/PDMS, carboxen/PDMS and polyacrylate, for autosampler useLog in for availability1 * 1 ST
- Kit 1: polyacrylate, PDMS, PDMS, for volatiles and semivolatiles, for manual useLog in for availability1 * 3 ST
- Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for manual useLog in for availability1 * 1 ST
- Kit 4: PDMS, PDMS/DVB and carboxen/PDMS, for flavors and odors, for manual useLog in for availability1 * 1 ST
- StableFlex fiber kit: PDMS/DVB, DVB/carboxen/PDMS, carboxen/PDMS and polyacrylate, for manual useLog in for availability1 * 1 ST
Accessories
Product Information
- DescriptionVerfügbarkeitPreis
- SPME link septa, 11 mmArtikel gestrichen-
Accessories
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
- VARI392609902Merlin microseal, SPME replacement seal, 23 gaugeLog in for availability1 * 1 ST - 439,89 €
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



