ContactUsLinkComponent
Foot adapter
Foot adapter
Catalog # RLAA2119
Supplier:  R L ENTERPRISES
Foot adapter
Catalog # RLAA2119
Supplier:  R L ENTERPRISES
Specifications
  • Description:
    Foot adapter, aluminium
  • Pk:
    1
  • For rod Ø:
    13 mm
Specifications
About this item
Adapter manufactured out of aluminium with boss to fit scaffold feet.
Accepts three 13 mm diameter rods or tubes at right angles (2 perpendicular to the boss and the third in line with the boss), with alien screws for fixing. One allen key provided per pack of ten.