Order Entry
Austria
ContactUsLinkComponent
 
DNA isolation, Genomic DNA isolation kits, XIT™
 
undefined
DNA isolation, Genomic DNA isolation kits, XIT™

 

  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    XIT™ DNA from blood

 

Frequently Bought Together