ContactUsLinkComponent
PCR tube rack with cover, SP Bel-Art
PCR tube rack with cover, SP Bel-Art
  BELAF189020005
undefined
PCR tube rack with cover, SP Bel-Art
  BELAF189020005
  • Colour:
    Assorted
  • Description:
    96-well PCR storage rack with cover
  • Seq:
    03
  • Pk:
    5
PP, with translucent PP cover.
Designed for preparing and storing 0,2 ml thin-walled PCR tubes and strip tubes before or after PCR sample processing applications. The compact rack features 96 wells that keep tubes securely upright in an 8×12 array. An alphanumeric grid assists in easy sample identification.
W×D×H: 127×89×34 mm
 : The Assortment pack includes one of each colour; flourescent blue, green, pink, yellow and orange.