ContactUsLinkComponent
Distilling bridge with Liebig condenser, Quickfit®
Distilling bridge with Liebig condenser, Quickfit®
Catalog # 201-0607
Supplier:  SciLabware
undefined
Distilling bridge with Liebig condenser, Quickfit®
Catalog # 201-0607
Supplier:  SciLabware
Specifications
  • Cone size:
    19/26 NS
  • Jacket length:
    155 mm
  • Socket size:
    14/23 NS
  • Pk:
    1
Specifications
About this item
PYREX®, borosilicate glass
  • Angle and position pre-defined
  • Less grease contamination
  • No air leakage under vacuum
  • No liquid hold-up at the joints
Comprises Liebig condenser, stillhead and receiver adapter in a single piece.