ContactUsLinkComponent
Adapters, receiver, vacuum, Quickfit®
Adapters, receiver, vacuum, Quickfit®
Catalog # BIBBRA11/SC
Supplier:  SciLabware
undefined
Adapters, receiver, vacuum, Quickfit®
Catalog # BIBBRA11/SC
Supplier:  SciLabware
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    19/26 NS
  • Pk:
    5
Specifications
About this item
PYREX®, borosilicate glass, bent.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Allows up to four flasks to be connected to a condenser outlet. Useful for collecting distillate fractions. Each flask can be progressively filled by rotating the adapter.